Learn More
Beckman Coulter Flow-Check Pro Fluorospheres
Catalog No. NC2399442
Encompass
Flow-Check Pro Fluorospheres is a suspension of fluorescent microspheres used for daily verification of the Navios™or FC 500 Flow Cytometer optical alignment and fluidics system.
This item is not returnable. View return policy
Catalog No. NC2399442 Supplier Beckman Coulter Supplier No. A63493
Flow-Check Pro Fluorospheres is a suspension of fluorescent microspheres used for daily verification of the Navios™or FC 500 Flow Cytometer optical alignment and fluidics system.