Learn More
MSC The 3M Portable Proportioner, 0.5 gal. Bottles, 1 ea
Catalog No. 502818358
Encompass
This flow control portable dispenser is designed for precise dispensing of chemical concentrates. It functions as a portable proportioner, accommodating one chemical product for convenient use.
  • Accommodates one chemical product
  • For use with 0.5 gal. chemical concentrate bottles
  • Portable design for flexible application
Catalog No. 50-281-8358 Supplier MSC Supplier No. 14162077
This flow control portable dispenser is designed for precise dispensing of chemical concentrates. It functions as a portable proportioner, accommodating one chemical product for convenient use.
  • Accommodates one chemical product
  • For use with 0.5 gal. chemical concentrate bottles
  • Portable design for flexible application