Learn More
MSC Flow Control System, Wall Mount, 1/Case
Catalog No. 502816253
Encompass
The Flow Control System Chemical Dispenser is a wall-mounted, compact, and lockable dispenser designed to safely and accurately dilute chemical concentrates to create ready-to-use cleaning products. It controls water flow by adjusting to changes in water pressures, ensuring an accurate ready-to-use (RTU) solution.
  • Offers 55% more accuracy at diluting chemical concentrates compared to competitive systems
  • Is 17% to 49% more compact than competitive four-bottle dispenser systems
  • Minimizes chemical waste
  • Improved cleaning chemical dilution reduces chemical use and waste
  • Easy to use chemical dispenser
  • Lockable chemical dispenser
Catalog No. 50-281-6253 Supplier MSC Supplier No. 14162127
The Flow Control System Chemical Dispenser is a wall-mounted, compact, and lockable dispenser designed to safely and accurately dilute chemical concentrates to create ready-to-use cleaning products. It controls water flow by adjusting to changes in water pressures, ensuring an accurate ready-to-use (RTU) solution.
  • Offers 55% more accuracy at diluting chemical concentrates compared to competitive systems
  • Is 17% to 49% more compact than competitive four-bottle dispenser systems
  • Minimizes chemical waste
  • Improved cleaning chemical dilution reduces chemical use and waste
  • Easy to use chemical dispenser
  • Lockable chemical dispenser