Learn More
MARVELGENT BIOSCIENCES INC Empty Disposable Gravity Flow Columns, 300 mL with 20 um frits, 10/pk with stopcocks
SDP
Catalog No. NC1804281
Encompass_Preferred
These disposable columns are economical, versatile columns for single-use purification applications using gravity flow. Simple, cost-effective, readily adaptable for a wide range of gravity flow applications. Made of high purity medical grade polypropylene, stable in commonly used chemicals and reagents. Available in various sizes to suit for different experiments.
Catalog No. NC1804281 Supplier MARVELGENT BIOSCIENCES INC Supplier No. 110261010SC
These disposable columns are economical, versatile columns for single-use purification applications using gravity flow. Simple, cost-effective, readily adaptable for a wide range of gravity flow applications. Made of high purity medical grade polypropylene, stable in commonly used chemicals and reagents. Available in various sizes to suit for different experiments.