Learn More
Chemglass Life Sciences Adapter, Flow Control, Chromatography, 45/50 Inner Joint
SDP
Catalog No. CG119102
Encompass_Preferred
Adapter, Flow Control, Chromatography, 45/50 Inner Joint. Flow control adapter for use with CG-1188 and CG-1189 chromatography columns along with CG-1190 reservoirs. Adapter has a 0-4mm Chem-Cap metering plug.
Catalog No. CG119102 Supplier Chemglass Life Sciences Supplier No. CG119102
Adapter, Flow Control, Chromatography, 45/50 Inner Joint. Flow control adapter for use with CG-1188 and CG-1189 chromatography columns along with CG-1190 reservoirs. Adapter has a 0-4mm Chem-Cap metering plug.