Learn More
Bellco Glass, Inc. Flow-Control Tube CLAMP,1/2OD
SDP
Catalog No. 501946383
Encompass_Preferred
Flow Control Tube Clamp ½". Plastic flow-control tube clamp. MADE IN THE USAMax.tube od: 1/2" (13mm)Max. Pressure: not ratedTemperature range: 32° to 200°fColor: white (opaque)Material: acetalSterilization: stem (autoclave)Materials meet: not ratedTubing: use with pvc, silicone, latex & rubber with a durometer of 35a-60aShut off clampPKG/QTY: 10
Catalog No. 50-194-6383 Supplier Bellco Glass, Inc. Supplier No. 780800004
Flow Control Tube Clamp ½". Plastic flow-control tube clamp. MADE IN THE USAMax.tube od: 1/2" (13mm)Max. Pressure: not ratedTemperature range: 32° to 200°fColor: white (opaque)Material: acetalSterilization: stem (autoclave)Materials meet: not ratedTubing: use with pvc, silicone, latex & rubber with a durometer of 35a-60aShut off clampPKG/QTY: 10