Learn More
Chemglass Life Sciences Tube Clamp, Flow Control, 12 Position, 0.45" OD
SDP
Catalog No. 50143759
Encompass_Preferred
Acetal clamp is autoclavable; won't rust, corrode or distort.Clamp accepts up to 0. 45" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.
Catalog No. 50-143-759 Supplier Chemglass Life Sciences Supplier No. CLS1436C01
Acetal clamp is autoclavable; won't rust, corrode or distort.Clamp accepts up to 0. 45" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.