Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Proportional Flowmeter Controller, Mass; 100 mL/min Gas
Catalog No. NC9555555
Encompass
Measure 30 standard gases while enjoying versatility and high accuracy from a laminar-based mass flow controller. Accuracy of 0.8% of reading 0.2% full-scale; repeatability of 0.2%. 100 to 1 turndown with ranges of 1 SCCM full-scale up to 1000 SLPM.
Catalog No. NC9555555 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3290759
Measure 30 standard gases while enjoying versatility and high accuracy from a laminar-based mass flow controller. Accuracy of 0.8% of reading 0.2% full-scale; repeatability of 0.2%. 100 to 1 turndown with ranges of 1 SCCM full-scale up to 1000 SLPM.