Learn More
Azzota Corp Flow cuvette 10mm 4x12 0.48ml
Catalog No. 5000887101
Encompass_Preferred
10mm Pathlength Flowthrough Cuvettes Inside width 4x12mm Volume - 0 48ml Optical Glass 340nm-2500nm
Catalog No. 50-008-87101 Supplier Azzota Corp Supplier No. G74
10mm Pathlength Flowthrough Cuvettes Inside width 4x12mm Volume - 0 48ml Optical Glass 340nm-2500nm