Learn More
Azzota Corp Flow cuvette 30mm 4x12 1.5ml g
Catalog No. 5000887019
Encompass_Preferred
Flowthrough Cuvette 30mm Pathlength 4x12mm Inside Width Optical Glass (340nm-2500nm) Volume - 1 5ml Dimensions - 45x12 5x32 5mm
Catalog No. 50-008-87019 Supplier Azzota Corp Supplier No. G76
Add to Cart
Add to Cart
Flowthrough Cuvette 30mm Pathlength 4x12mm Inside Width Optical Glass (340nm-2500nm) Volume - 1 5ml Dimensions - 45x12 5x32 5mm