Learn More
Raybiotech Inc Flow Cytometry Compensation Beads, 100 Tests
Catalog No. NC3940328
Encompass_Preferred
The RayBright® Flow Cytometry Compensation Beads are designed to capture all isotypes of common primary antibodies to allow for easy and rapid created of FACS compensation beads. Our beads can capture all rat, mouse, rabbit, Syrian Hamster and Armenian hamster antibodies that are labeled with your fluorophores of interest. RayBright® Flow Cytometry Compensation Beads are made up of two populations: negative beads and positive beads, with only the positive beads binding the fluorochrome-conjugated antibodies. It’s easy and convenient to use, and a perfect tool for multi-color flow cytometry compensation.
Catalog No. NC3940328 Supplier Raybiotech Inc Supplier No. 13700013100
The RayBright® Flow Cytometry Compensation Beads are designed to capture all isotypes of common primary antibodies to allow for easy and rapid created of FACS compensation beads. Our beads can capture all rat, mouse, rabbit, Syrian Hamster and Armenian hamster antibodies that are labeled with your fluorophores of interest. RayBright® Flow Cytometry Compensation Beads are made up of two populations: negative beads and positive beads, with only the positive beads binding the fluorochrome-conjugated antibodies. It’s easy and convenient to use, and a perfect tool for multi-color flow cytometry compensation.