Learn More
Abcam Flow Cytometry Fi x ation & Permeabilization Kit, 50 TestsS
Catalog No. 502699006
Encompass_Preferred
Flow Cytometry Fixation & Permeabilization Kit ab314689 contains optimally formulated buffers for fixation and permeabilization of cells for immunofluorescence staining of intracellular antigens for analysis by flow cytometry.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-9006 Supplier Abcam Supplier No. AB31468950TESTS
Flow Cytometry Fixation & Permeabilization Kit ab314689 contains optimally formulated buffers for fixation and permeabilization of cells for immunofluorescence staining of intracellular antigens for analysis by flow cytometry.
The product is subject to the following: Abcam Restricted Use Statement