Learn More
Azzota Corp Flow Fluor 10mm 0.48ml Qz
Catalog No. 5000887179
Encompass_Preferred
Self masking continuous flowthrough Fluorometer cell 10mm Pathlength window size 4x12mm 0 48ml Quartz 190-2500nm
Catalog No. 50-008-87179 Supplier Azzota Corp Supplier No. Q74F
Self masking continuous flowthrough Fluorometer cell 10mm Pathlength window size 4x12mm 0 48ml Quartz 190-2500nm