Learn More
BURKLE INC Flow indicator CheMobil, PMP, red
Catalog No. 5000026101
Encompass_Preferred
Flow indicators give precise visual indication of the flow velocity of liquids or gases being transported through hose connections. The flow indicator with impeller functions in both directions and in any position. The impeller (PP) starts to rotate at a flow rate about 150 ml/min (5.07 oz./min) (liquids).
Catalog No. 50-000-26101 Supplier BURKLE INC Supplier No. 87202002
Flow indicators give precise visual indication of the flow velocity of liquids or gases being transported through hose connections. The flow indicator with impeller functions in both directions and in any position. The impeller (PP) starts to rotate at a flow rate about 150 ml/min (5.07 oz./min) (liquids).