Learn More
BURKLE INC Flow indicator LiquiMobil, SAN, red
Catalog No. NC2187883
Encompass_Preferred
Flow indicators give precise visual indication of the flow velocity of liquids or gases being transported through hose connections. The flow indicator with impeller functions in both directions and in any position. The impeller (PP) starts to rotate at a flow rate about 150 ml/min (5.07 oz./min) (liquids).
This item is not returnable. View return policy
Catalog No. NC2187883 Supplier BURKLE INC Supplier No. 87201002
Flow indicators give precise visual indication of the flow velocity of liquids or gases being transported through hose connections. The flow indicator with impeller functions in both directions and in any position. The impeller (PP) starts to rotate at a flow rate about 150 ml/min (5.07 oz./min) (liquids).