Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area Flowmeter with Valve, Acrylic Housing and Brass Fittings, 50 mm Scale, 0.1 LPM Air
Catalog No. NC9804598
Encompass
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters¿ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately) which allows for mobility from bench to bench.
Catalog No. NC9804598 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3246042
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters¿ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately) which allows for mobility from bench to bench.