Learn More
STEMCELL Technologies Single Flow Meter, Size: 1 Unit
SDP
Catalog No. NC9972081
Encompass
Single Flow Meter allows for precise control of gas flow to generate a hypoxic environment for tissue culture using the Hypoxia Incubator Chamber (Catalog #27310). Features of the Single Flow Meter include: • Same precision as the flow meter used for full size continuous flow carbon dioxide incubator chambers• Calibrated for 5 - 50 liters per minute (LPM)
Catalog No. NC9972081 Supplier STEMCELL Technologies Supplier No. 27311
Single Flow Meter allows for precise control of gas flow to generate a hypoxic environment for tissue culture using the Hypoxia Incubator Chamber (Catalog #27310). Features of the Single Flow Meter include: • Same precision as the flow meter used for full size continuous flow carbon dioxide incubator chambers• Calibrated for 5 - 50 liters per minute (LPM)