Learn More
Grainger Gems Sensors and Controls RotorFlow Visual Indicator, Brass, 1/4 in. FNPT, 0.5 to 5.0 GPM, 1 ea.
Catalog No. 502794098
Encompass_Preferred
These visual indicators provide a reliable, simple method for monitoring fluid flow. Using a bright orange rotor that turns in response to flow, they allow users to visually estimate or confirm flow rates. These indicators feature a versatile design where either port can accommodate incoming flow, and the bayonet-mounting lens is easily removed for rapid maintenance and cleaning.
  • Provides clear, visual flow rate confirmation
  • Features a durable brass body construction
  • Equipped with a quick-clean, bayonet-mounting lens
  • Allows installation with either port for incoming flow
  • Designed for straightforward and simple operation
  • Operating temperature range of -29°C to 100°C (-20°F to 212°F)
Catalog No. 50-279-4098 Supplier Grainger Supplier No. 1AMD7
These visual indicators provide a reliable, simple method for monitoring fluid flow. Using a bright orange rotor that turns in response to flow, they allow users to visually estimate or confirm flow rates. These indicators feature a versatile design where either port can accommodate incoming flow, and the bayonet-mounting lens is easily removed for rapid maintenance and cleaning.
  • Provides clear, visual flow rate confirmation
  • Features a durable brass body construction
  • Equipped with a quick-clean, bayonet-mounting lens
  • Allows installation with either port for incoming flow
  • Designed for straightforward and simple operation
  • Operating temperature range of -29°C to 100°C (-20°F to 212°F)