Learn More
Grainger Gems Sensors RFI Series Rotor Flow Indicator, Brass, 1 in FNPT, 8-60 GPM, 1 ea
Catalog No. 502793591
Encompass_Preferred
The RFI Series Rotorflow is an inline visual flow indicator designed to provide accurate flow monitoring by using a rotating impeller to represent media movement. This device features a rugged brass housing and a bright, spinning rotor for clear visual confirmation of liquid flow in a system.
  • Flow range of 8.0 to 60.0 GPM
  • Maximum pressure rating of 200 psi
  • Fluid temperature range of -20 °F to +212 °F
  • Durable brass body with a polysulfone lens that is easily removed for quick cleanout
  • Ceramic rotor pin for reliable operation in harsh conditions
  • 1 in. FNPT connection size
  • UL Recognized, CE, and CUL recognized approvals
  • RoHS compliant
Catalog No. 50-279-3591 Supplier Grainger Supplier No. 1AME1
The RFI Series Rotorflow is an inline visual flow indicator designed to provide accurate flow monitoring by using a rotating impeller to represent media movement. This device features a rugged brass housing and a bright, spinning rotor for clear visual confirmation of liquid flow in a system.
  • Flow range of 8.0 to 60.0 GPM
  • Maximum pressure rating of 200 psi
  • Fluid temperature range of -20 °F to +212 °F
  • Durable brass body with a polysulfone lens that is easily removed for quick cleanout
  • Ceramic rotor pin for reliable operation in harsh conditions
  • 1 in. FNPT connection size
  • UL Recognized, CE, and CUL recognized approvals
  • RoHS compliant