Learn More
Grainger Gems Sensors RFO RotorFlow Electronic Flow Sensor, Brass, 0.5-5 GPM, 1/4 in. FNPT, 1 ea
Catalog No. 502794073
Encompass_Preferred
The RFO series electronic flow sensor is a durable rotary flow switch engineered for hydraulic and lubrication systems. It provides dependable flow monitoring in demanding industrial environments by utilizing high-visibility rotors paired with solid-state electronics to deliver reliable pulsed output signals.
  • Features a brass housing rated for pressures up to 200 psi.
  • Supports a measurement range of 0.5 to 5.0 GPM with ±7% accuracy.
  • Operates within a temperature range of -20°F to +212°F.
  • Includes a 24 in. long, 22 AWG, 3 conductor PVC cable for secure termination.
  • Utilizes a 1/4 in. FNPT connection and includes a low flow adapter.
  • Ideal for water purification, semiconductor processing equipment, chillers, and lasers.
Catalog No. 50-279-4073 Supplier Grainger Supplier No. 1AMD1
The RFO series electronic flow sensor is a durable rotary flow switch engineered for hydraulic and lubrication systems. It provides dependable flow monitoring in demanding industrial environments by utilizing high-visibility rotors paired with solid-state electronics to deliver reliable pulsed output signals.
  • Features a brass housing rated for pressures up to 200 psi.
  • Supports a measurement range of 0.5 to 5.0 GPM with ±7% accuracy.
  • Operates within a temperature range of -20°F to +212°F.
  • Includes a 24 in. long, 22 AWG, 3 conductor PVC cable for secure termination.
  • Utilizes a 1/4 in. FNPT connection and includes a low flow adapter.
  • Ideal for water purification, semiconductor processing equipment, chillers, and lasers.