Learn More
Grainger Gems Sensors RFO Series Flow Rate Monitor, Polypropylene, 0.5-5 GPM, 1/4 in. FNPT, 1 ea.
Catalog No. 502794064
Encompass_Preferred
The RFO Series Electronic Flow Sensor is a durable rotary flow switch designed for monitoring hydraulic and lubrication systems in industrial environments. These sensors integrate high-visibility rotors with solid-state electronics in a compact, panel-mountable housing, making them suitable for applications such as semiconductor processing equipment, water purification, and chillers.
  • Provides reliable flow monitoring with a flow rate range of 0.5 GPM to 5 GPM.
  • Features a durable polypropylene body with Buna N O-ring seals.
  • Includes a low flow adapter for versatile installation.
  • Designed for operating pressures up to 100 psi and fluid temperatures from -20°F to +180°F.
  • Outputs 2,200 pulses per gallon for precise measurement.
  • Equipped with a polysulfone sight window for easy monitoring.
Catalog No. 50-279-4064 Supplier Grainger Supplier No. 1AMB8
The RFO Series Electronic Flow Sensor is a durable rotary flow switch designed for monitoring hydraulic and lubrication systems in industrial environments. These sensors integrate high-visibility rotors with solid-state electronics in a compact, panel-mountable housing, making them suitable for applications such as semiconductor processing equipment, water purification, and chillers.
  • Provides reliable flow monitoring with a flow rate range of 0.5 GPM to 5 GPM.
  • Features a durable polypropylene body with Buna N O-ring seals.
  • Includes a low flow adapter for versatile installation.
  • Designed for operating pressures up to 100 psi and fluid temperatures from -20°F to +180°F.
  • Outputs 2,200 pulses per gallon for precise measurement.
  • Equipped with a polysulfone sight window for easy monitoring.