Learn More
Grainger Gems Sensors RFO RotorFlow Electronic Flow Sensor, Brass, 1 NPT, 1 ea
Catalog No. 502794062
Encompass_Preferred
The RotorFlow electronic flow sensor combines a high-visibility paddlewheel design with integrated solid-state electronics in a compact, panel-mountable housing. Designed for reliable flow rate monitoring, these units utilize Hall effect technology to provide a pulsed voltage output proportional to flow, suitable for diverse industrial applications including water purification, semiconductor processing, and thermal management systems.
  • Provides accurate pulsed VDC flow rate output with visual confirmation.
  • Features brass body construction with a durable PPS composite rotor and ceramic rotor pin.
  • Operates with input voltages from 4.5 VDC to 24 VDC.
  • Supports flow rates from 8 GPM to 60 GPM with a frequency output range of 15 Hz to 225 Hz.
  • Maximum operating pressure of 200 psig.
  • Compatible with standard digital logic families.
Catalog No. 50-279-4062 Supplier Grainger Supplier No. 1AMD4
The RotorFlow electronic flow sensor combines a high-visibility paddlewheel design with integrated solid-state electronics in a compact, panel-mountable housing. Designed for reliable flow rate monitoring, these units utilize Hall effect technology to provide a pulsed voltage output proportional to flow, suitable for diverse industrial applications including water purification, semiconductor processing, and thermal management systems.
  • Provides accurate pulsed VDC flow rate output with visual confirmation.
  • Features brass body construction with a durable PPS composite rotor and ceramic rotor pin.
  • Operates with input voltages from 4.5 VDC to 24 VDC.
  • Supports flow rates from 8 GPM to 60 GPM with a frequency output range of 15 Hz to 225 Hz.
  • Maximum operating pressure of 200 psig.
  • Compatible with standard digital logic families.