Learn More
Warner Instruments™ Flow Regulating Valve, with On/Off Switch
Catalog No. BTX640221 Shop All BTX Harvard Products
Encompass_Preferred
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
BTX640221 Stainless Steel
1 options
Catalog No. BTX640221 Supplier Warner Instruments™ Supplier No. 640221
Flow-regulating valve with an on/off switch provides fine solution control in gravity-fed perfusion or vacuum applications.
  • Reproducible settings
  • Mates with standard PE-160 tubing
  • Available with on/off switch
Specifications
For Use With (Application) Flow Regulation
For Use With PE-160 Tubing
Material Stainless Steel
Model FR-55S
Type Fine Control Valve