Learn More
Thermo Scientific Flow Sensor
Catalog No. NC0324212 Shop All Thermo Scientific Products
Encompass
A flow sensor measuring fluid flow rate by spinning rotor and electronics translating rotor speed to output signal proportional to flow rate, used for monitoring and controlling fluid flow in industrial or scientific applications.
Catalog No. NC0324212 Supplier Thermo Scientific Supplier No. 1191740
A flow sensor measuring fluid flow rate by spinning rotor and electronics translating rotor speed to output signal proportional to flow rate, used for monitoring and controlling fluid flow in industrial or scientific applications.