Learn More
Hach Company Flow Sensor for TU5300/5400
Catalog No. 5000207726 Shop All Hach Company Products
Encompass
Flow Sensor for TU5300sc and TU5400sc Laser TurbidimeterAdding a flow measurement along with your TU5300sc or TU5400sc turbidity measurement provides critical information about the status of your drinking water process A TU5 series turbidimeter s flow sensor accessory can provide alerts to flow issues giving you important information to resolve problems and get your plant running smoothly
Catalog No. 50-002-07726 Supplier Hach Company Supplier No. LQV160.99.00002
Flow Sensor for TU5300sc and TU5400sc Laser TurbidimeterAdding a flow measurement along with your TU5300sc or TU5400sc turbidity measurement provides critical information about the status of your drinking water process A TU5 series turbidimeter s flow sensor accessory can provide alerts to flow issues giving you important information to resolve problems and get your plant running smoothly