Learn More
Chemglass Life Sciences FlowTubes with Standard Cap, Sterile, 12 x 75mm, 25/bag
SDP
Catalog No. 501979488
Encompass_Preferred
FlowTubes with Standard Cap, Sterile, 12 x 75mm, 25/bag. Made of crystal-clear polystyrene, these 12 x 75mm FlowTubes™ are designed for Fluorescence Activated Cell Sorting/ flow cytometry equipment including FACS systems from BD Biosciences. The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with the automated instrument. These tubes are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments. Available with standard two-position culture cap.
Catalog No. 50-197-9488 Supplier Chemglass Life Sciences Supplier No. CLS4380010
FlowTubes with Standard Cap, Sterile, 12 x 75mm, 25/bag. Made of crystal-clear polystyrene, these 12 x 75mm FlowTubes™ are designed for Fluorescence Activated Cell Sorting/ flow cytometry equipment including FACS systems from BD Biosciences. The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with the automated instrument. These tubes are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments. Available with standard two-position culture cap.