Learn More
Chemglass Life Sciences FlowTubes with Strainer Cap, 12 x 75mm, Sterile, Individually Wrapped
SDP
Catalog No. 501979486
Encompass_Preferred
FlowTubes with Strainer Cap, 12 x 75mm, Sterile, Individually Wrapped. Made of crystal-clear polystyrene, these 12 x 75mm FlowTubes™ are designed for Fluorescence Activated Cell Sorting/ flow cytometry equipment including FACS systems from BD Biosciences. The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with the automated instrument. These tubes are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments. Cap contains a 35µm nylon mesh screen, designed to assist in cell sorting/straining applications.
Catalog No. 50-197-9486 Supplier Chemglass Life Sciences Supplier No. CLS4380007
FlowTubes with Strainer Cap, 12 x 75mm, Sterile, Individually Wrapped. Made of crystal-clear polystyrene, these 12 x 75mm FlowTubes™ are designed for Fluorescence Activated Cell Sorting/ flow cytometry equipment including FACS systems from BD Biosciences. The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with the automated instrument. These tubes are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments. Cap contains a 35µm nylon mesh screen, designed to assist in cell sorting/straining applications.