Learn More
Hach Company Flow-Thru CPVC Mt. Hard. Tee
Supplier:  Hach Company MH336
Encompass
Mounting Hardware-Flow-Thru-CPVC-1-1/2 InchCPVC 1-1/2 Inch Tee and Aluminum Junction Box for Use with LCP pH/ORP Sensors
Catalog No. 50-002-07257