Learn More
MSC Dow Flowable Adhesive Sealant, RTV Silicone, Clear, MIL-A-46106, 90 mL
Catalog No. 501149236
Encompass
This one-part, solventless silicone elastomer is designed for general sealing and bonding applications. It is a clear RTV silicone joint sealant suitable for various laboratory and industrial uses, adhering effectively to glass, plastic, and metal.
  • One-part, solventless silicone elastomer formulation
  • Clear RTV silicone joint sealant
  • Full cure time of 24 hours
  • Tack-free dry time of 13 minutes
  • Operating temperature range from -65.0 °C to 180 °C
  • Adheres to glass, plastic, and metal surfaces
  • Meets MIL-A-46106 standards for quality
Catalog No. 50-114-9236 Supplier MSC Supplier No. 00232181
This one-part, solventless silicone elastomer is designed for general sealing and bonding applications. It is a clear RTV silicone joint sealant suitable for various laboratory and industrial uses, adhering effectively to glass, plastic, and metal.
  • One-part, solventless silicone elastomer formulation
  • Clear RTV silicone joint sealant
  • Full cure time of 24 hours
  • Tack-free dry time of 13 minutes
  • Operating temperature range from -65.0 °C to 180 °C
  • Adheres to glass, plastic, and metal surfaces
  • Meets MIL-A-46106 standards for quality