Learn More
Waters Corp Flowcell, Autopurification, 2487/2489 Series, 2000 psi Max Pressure
Catalog No. 502390583 Shop All Waters Corporation Products
Encompass_Preferred
This autopurification flowcell is designed for use with 2487 and 2489 UV/Visible Detectors, providing reliable performance for demanding laboratory applications.
  • Compatible with 2487/2489 series detectors
  • Maximum pressure rating of 2000 psi
Catalog No. 50-239-0583 Supplier Waters Corp Supplier No. 289000614
This autopurification flowcell is designed for use with 2487 and 2489 UV/Visible Detectors, providing reliable performance for demanding laboratory applications.
  • Compatible with 2487/2489 series detectors
  • Maximum pressure rating of 2000 psi