Learn More
Waters Corp Waters Corporation Flowcell, Low Flow, ACQUITY TUV
Catalog No. 50791706 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters ACQUITY TUV Flowcell is a critical component for high-performance liquid chromatography systems, integrating advanced light-guiding technology to deliver precise and sensitive detection. This flowcell is engineered for robust performance in demanding laboratory environments.
  • Features a light-guiding UPLC flow cell design.
  • Incorporates Intelligent Chip Technology for enhanced performance.
  • Ensures precise and sensitive detection of analytes.
  • Common wetted materials include 316 stainless steel, fused silica, Teflon™ AF, and PEEK.
  • Utilizes total internal reflection, analogous to an optical fiber, for efficient light transmission through the sample.
  • Facilitates sample introduction and removal via PEEK tubing.
Catalog No. 50-791-706 Supplier Waters Corp Supplier No. 205015001
Add to Cart
Add to Cart
Request a Quote
The Waters ACQUITY TUV Flowcell is a critical component for high-performance liquid chromatography systems, integrating advanced light-guiding technology to deliver precise and sensitive detection. This flowcell is engineered for robust performance in demanding laboratory environments.
  • Features a light-guiding UPLC flow cell design.
  • Incorporates Intelligent Chip Technology for enhanced performance.
  • Ensures precise and sensitive detection of analytes.
  • Common wetted materials include 316 stainless steel, fused silica, Teflon™ AF, and PEEK.
  • Utilizes total internal reflection, analogous to an optical fiber, for efficient light transmission through the sample.
  • Facilitates sample introduction and removal via PEEK tubing.