Learn More
Waters Corp Flowcell Rebuild Kit, 2998 Autopurification, HPLC
Catalog No. 502390685 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit contains the lenses and gaskets necessary to rebuild a 2998 autopurification flow cell.
  • Contains the necessary lenses and gaskets.
  • Designed for rebuilding 2998 autopurification flow cells.
  • Compatible with HPLC systems.
  • Replaced as required to maintain optimal performance.
  • Supplied as a single pack.
Catalog No. 50-239-0685 Supplier Waters Corp Supplier No. 700005102
This rebuild kit contains the lenses and gaskets necessary to rebuild a 2998 autopurification flow cell.
  • Contains the necessary lenses and gaskets.
  • Designed for rebuilding 2998 autopurification flow cells.
  • Compatible with HPLC systems.
  • Replaced as required to maintain optimal performance.
  • Supplied as a single pack.