Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLINGSHOT BIOSCIENCES FlowCytes WBC (Fixed), 25 tests
SDP

Catalog No. NC2127317
Encompass_Preferred

FlowCytes WBC (Fixed) are a synthetic polymer based cell mimic with defined scatter populations designed to imitate fixed lymphocyte, monocyte and granulocyte populations to support standardization and reproducibility of scatter settings across assays.

Specifications:
Catalog Number: SSB-15-A
Package Size: 25 tests
Storage Conditions: 2 - 30°C
Shipping Conditions: No Ice
Shelf Life: 36 months
Country of Origin: United States
For Research Use Only.

This item is not returnable. View return policy
Catalog No. NC2127317 Supplier SLINGSHOT BIOSCIENCES Supplier No. SSB15A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

FlowCytes WBC (Fixed) are a synthetic polymer based cell mimic with defined scatter populations designed to imitate fixed lymphocyte, monocyte and granulocyte populations to support standardization and reproducibility of scatter settings across assays.

Specifications:
Catalog Number: SSB-15-A
Package Size: 25 tests
Storage Conditions: 2 - 30°C
Shipping Conditions: No Ice
Shelf Life: 36 months
Country of Origin: United States
For Research Use Only.

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.