Learn More
Beckman Coulter Flowerplate Offline Cultivation 48 EA
Catalog No. NC1728262
Encompass
48-well microplate for cultivating cells offline in laboratory with standard well spacing and depth for multichannel pipettes, automated liquid handlers, and plate readers; allows offline cellular analysis without cytometer; use with Kaluza software for analyzing data
Catalog No. NC1728262 Supplier Beckman Coulter Supplier No. M2PMTP48OFF
48-well microplate for cultivating cells offline in laboratory with standard well spacing and depth for multichannel pipettes, automated liquid handlers, and plate readers; allows offline cellular analysis without cytometer; use with Kaluza software for analyzing data