Learn More
Beckman Coulter Beckman Coulter M2P-MTP-48-B FlowerPlate 48-Well Microtiter Plate, Without Optodes
Catalog No. NC1422056
Encompass
The Beckman Coulter M2P-MTP-48-B FlowerPlate is a 48-well microtiter plate in the standard ANSI/SLAS (SBS) format designed to improve gas transfer and enable online monitoring in microbioreactor workflows. Its flower-shaped, baffled wells enhance aeration and mixing; this variant is supplied without integrated optical sensors (optodes) and is intended for use with compatible microbioreactor readout systems.
  • 48-well microtiter plate.
  • Standard ANSI/SLAS (SBS) footprint.
  • Flower-shaped wells to promote aeration and mixing.
  • Supplied without optical sensors (no optodes).
  • Enables online biomass and fluorescence readout with compatible microbioreactor systems.
  • Optimized for increased gas transfer in small-scale cultivations.
Catalog No. NC1422056 Supplier Beckman Coulter Supplier No. M2PMTP48B
The Beckman Coulter M2P-MTP-48-B FlowerPlate is a 48-well microtiter plate in the standard ANSI/SLAS (SBS) format designed to improve gas transfer and enable online monitoring in microbioreactor workflows. Its flower-shaped, baffled wells enhance aeration and mixing; this variant is supplied without integrated optical sensors (optodes) and is intended for use with compatible microbioreactor readout systems.
  • 48-well microtiter plate.
  • Standard ANSI/SLAS (SBS) footprint.
  • Flower-shaped wells to promote aeration and mixing.
  • Supplied without optical sensors (no optodes).
  • Enables online biomass and fluorescence readout with compatible microbioreactor systems.
  • Optimized for increased gas transfer in small-scale cultivations.