Learn More
Electron Microscopy Sciences FlowMesh Sheet
SDP
Catalog No. 5034222
Encompass_Preferred
A Permeable Support Mesh for Gels and Membranes Protects - Place FlowMesh above or below a gel or filter to protect from tearing, bending or folding
Sturdy - Supports gels during incubation with nucleic acid probes, antibody probes, equilibration buffers, stain, destain solutions, etc.
Versatile - Use as a permeable spacer for stacking as well as for staining and destaining procedures
Access - Both sides of a container or plastic bag are assured of constant solution access, which improves the uniformity of hybridizations and binding assays.
Convenient - FlowMesh is re-usable and easily cut to size
Size: 8.50" x 11.00"
Catalog No. 50-342-22 Supplier Electron Microscopy Sciences Supplier No. 6850520
A Permeable Support Mesh for Gels and Membranes Protects - Place FlowMesh above or below a gel or filter to protect from tearing, bending or folding
Sturdy - Supports gels during incubation with nucleic acid probes, antibody probes, equilibration buffers, stain, destain solutions, etc.
Versatile - Use as a permeable spacer for stacking as well as for staining and destaining procedures
Access - Both sides of a container or plastic bag are assured of constant solution access, which improves the uniformity of hybridizations and binding assays.
Convenient - FlowMesh is re-usable and easily cut to size
Size: 8.50" x 11.00"