Learn More
Hach Company Flowmeter, Air
Supplier:  Hach Company 1787500
Encompass
Styrene acrylonitrile plastic flowmeter measures air flow in the range of 0.2 to 2.0 SCFH. Used with the Cyanide Distillation Apparatus Set. Dwyer Instrument.
Catalog No. 50-199-5166