Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable Area Flowmeter with High-Res Valve, Correlated Reading, High-Flow, Aluminum Fittings; 65-mm Scale; 42.094 LPM Air
Catalog No. 50627220
Encompass
Aluminum is economical and good for general use with noncorrosive gases and liquids. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
Catalog No. 50-627-220 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3204438
Aluminum is economical and good for general use with noncorrosive gases and liquids. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).