Learn More
Thermo Scientific FLOWMETER BIOCOMP W/NANO FLOW
Catalog No. NC2543359 Shop All Thermo Scientific Products
Encompass
3000 RSLC Nano System Flow Meters; flowmeter w/ nano flow selector; For Use With (Equipment): Capillary LC (0.5 to 10 uL/Min)
Catalog No. NC2543359 Supplier Thermo Scientific Supplier No. 6041.7902A
Add to Cart
Add to Cart
Request a Quote
3000 RSLC Nano System Flow Meters; flowmeter w/ nano flow selector; For Use With (Equipment): Capillary LC (0.5 to 10 uL/Min)