Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area Flowmeter, Correlated Reading, Brass Housing and Fittings, 65-mm Scale, 299 mL/min Air
Catalog No. 501961123
Encompass
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility.
Catalog No. 50-196-1123 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0326860
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility.