Learn More
Chemglass Life Sciences Nitrogen Flowmeter, 1.0 to 10L/min
SDP
Catalog No. 50872100
Encompass_Preferred
Nitrogen Flowmeter, 1.0 to 10L/min. Precision machined clear acrylic Nitrogen flowmeter with Brass metering valve is for use with Chemglass process reactors. Front of flowmeter has a blue scale indicating the L/min of air. HDPE hose connection is for use with 5/16” ID tubing. Unit is supplied complete with a 1/2” OD mounting arm for ease in clamping to support frame. Flowmeters with larger L/min ranges are available. Please contact us for details.
Catalog No. 50-872-100 Supplier Chemglass Life Sciences Supplier No. CG1605A14
Nitrogen Flowmeter, 1.0 to 10L/min. Precision machined clear acrylic Nitrogen flowmeter with Brass metering valve is for use with Chemglass process reactors. Front of flowmeter has a blue scale indicating the L/min of air. HDPE hose connection is for use with 5/16” ID tubing. Unit is supplied complete with a 1/2” OD mounting arm for ease in clamping to support frame. Flowmeters with larger L/min ranges are available. Please contact us for details.