Learn More
Wilmad Labglass FLOWMETER SOAP FILM 10ML
Catalog No. 50804209
Encompass
Flow Meter, Soapfilm. Capacity: 10mL; Subdivision: 0.1mL. Used to measure flow rate of carrier gas in gas chromatography. Supplied w/ LG-4340-100 rubber bulb.
Catalog No. 50-804-209 Supplier Wilmad Labglass Supplier No. LG8612095
Flow Meter, Soapfilm. Capacity: 10mL; Subdivision: 0.1mL. Used to measure flow rate of carrier gas in gas chromatography. Supplied w/ LG-4340-100 rubber bulb.