Learn More
Research Products International Corp FLOWMI Tip Strainer 40um 50cs
SDP
Catalog No. 502932457
Encompass
FLOWMI Tip Strainers remove large particles from a liquid sample prior to performing FACS and FLOW analysis. Features a 40um pore size for convenient and efficient filtering of small volumes up to 1000ul. FLOWMI strainers fit most brand of 1000ul pipette tips. Supplied sterile in a compact rack with a convenient sliding lid. Easy to use - simply attach by pressing the loaded pipette tip into the strainer then dispense sample. Sterility of unused strainers can be maintained by opening and re-sealing rack under a Laminar flow hood.
Catalog No. 50-293-2457 Supplier Research Products International Corp Supplier No. 246800
FLOWMI Tip Strainers remove large particles from a liquid sample prior to performing FACS and FLOW analysis. Features a 40um pore size for convenient and efficient filtering of small volumes up to 1000ul. FLOWMI strainers fit most brand of 1000ul pipette tips. Supplied sterile in a compact rack with a convenient sliding lid. Easy to use - simply attach by pressing the loaded pipette tip into the strainer then dispense sample. Sterility of unused strainers can be maintained by opening and re-sealing rack under a Laminar flow hood.