Learn More
Sigma Aldrich Fine Chemicals Biosciences flowmiR Cell Strainers por
Catalog No. 502900061
Encompass
Scienceware(R) flowmi Cell Strainers provide fast and efficient filtering of small volume samples (up to 1000mul) prior to fLOW or Analysis. Patent pending.
Catalog No. 50-290-0061 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BAH13680007050EA
Scienceware(R) flowmi Cell Strainers provide fast and efficient filtering of small volume samples (up to 1000mul) prior to fLOW or Analysis. Patent pending.