Learn More
Sigma Aldrich Fine Chemicals Biosciences flowmiR Cell Strainers por
Supplier:  Sigma Aldrich Fine Chemicals Biosciences BAH13680004050EA
Encompass
Scienceware(R) flowmi Cell Strainers provide fast and efficient filtering of small volume samples (up to 1000mul) prior to fLOW or Analysis. Patent pending.
Catalog No. 50-290-1338