Learn More
AVANTOR MASTERFLEX (VWR LLC) FLOWMTR ACRL NI 1.3 LPM 1/8
Catalog No. NC1945676
Encompass
Masterflex Variable-Area Flowmeter, Direct Reading, Acrylic Housing and Stainless Steel Fittings without valve, 1.3 L/min Nitrogen
Catalog No. NC1945676 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX6856063
Masterflex Variable-Area Flowmeter, Direct Reading, Acrylic Housing and Stainless Steel Fittings without valve, 1.3 L/min Nitrogen