Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area Flowtube, Correlated Reading, 65-mm Scale; 2,040 mL/min Air and 39.7 mL/min Water
Catalog No. 50627119
Encompass
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs. Choose 65- or 150-mm flowtubes depending on the selected frame size. Flowtubes are calibrated for direct-reading or correlated reading of air or water at STP (standard temperature and pressure).
Catalog No. 50-627-119 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0321958
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs. Choose 65- or 150-mm flowtubes depending on the selected frame size. Flowtubes are calibrated for direct-reading or correlated reading of air or water at STP (standard temperature and pressure).