Learn More
Electron Microscopy Sciences FLOWTUBE W/STRNR CAP IND WRP
SDP
Catalog No. 502413022
Encompass_Preferred
The FlowTubes for FACS are made of crystal clear virgin polystyrene, these 12x75mm FACS tubes are designed for most Flourescence Activated Cell Sorting/flow cytometry equipment, including FACS systems from 8D Biosciences. The diameter and wall thickness of the tube's neck are held to a tight tolerances to ensure consistent compatibility with automated instrumentation. They are available with the standard two-position culture cap, as well as with a cap that contains a 35µm nylon mesh screen, designed to assist in cell sorting/straining applications. These tubes and mesh caps are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments.
Catalog No. 50-241-3022 Supplier Electron Microscopy Sciences Supplier No. 6463900
The FlowTubes for FACS are made of crystal clear virgin polystyrene, these 12x75mm FACS tubes are designed for most Flourescence Activated Cell Sorting/flow cytometry equipment, including FACS systems from 8D Biosciences. The diameter and wall thickness of the tube's neck are held to a tight tolerances to ensure consistent compatibility with automated instrumentation. They are available with the standard two-position culture cap, as well as with a cap that contains a 35µm nylon mesh screen, designed to assist in cell sorting/straining applications. These tubes and mesh caps are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments.