Learn More
Chemglass Life Sciences FlowTubes without Cap, Non-Sterile, Bulk Econo-Pack, 4000/Case
SDP
Catalog No. 501979491
Encompass_Preferred
FlowTubes without Cap, Non-Sterile, Bulk Econo-Pack, 4000/Case. Made of crystal-clear polystyrene, these 12 x 75mm FlowTubes™ are designed for Fluorescence Activated Cell Sorting / flow cytometry equipment including FACS systems from BD Biosciences. The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with the automated instrument. These tubes are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments.
Catalog No. 50-197-9491 Supplier Chemglass Life Sciences Supplier No. CLS4380020
FlowTubes without Cap, Non-Sterile, Bulk Econo-Pack, 4000/Case. Made of crystal-clear polystyrene, these 12 x 75mm FlowTubes™ are designed for Fluorescence Activated Cell Sorting / flow cytometry equipment including FACS systems from BD Biosciences. The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with the automated instrument. These tubes are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments.