Learn More
Electron Microscopy Sciences Microscopic Fluid Chips - Standard
SDP
Supplier:  Electron Microscopy Sciences FV105
Encompass
FlowView starter kit provides world-leading innovative liquid sample inspection with SEM. These economic and easy-to-use starter kits include stage, accessories, and 24 Microscopic Fluid Chips. The chips are compatible with SEM for most major brands. Kit contains tweezer, pipette, torque wrench, carrier box, storage box, and 24 microscopic fluid chips. Microscopic fluid chips for use with most sample droplets (paint, mixing slurry, particles in oil).
Catalog No. 50-192-8105